Learn More
Alkali Scientific ASI™ Agarose LM, Low Melt, 100g
SDP
Catalog No. 502389573
Encompass_Preferred
ASI™ Agarose LM is a low melting point agarose ideal for preparative gels. The melting and gelling points are well below that of the DNA/RNA denaturation temperatures, allowing for recovery of nucleotides from the gel. Also useful for cloning tissue culture cells and viral plaque assays. All of our agarose is manufactured under a “green” process that eliminates the use of organic solvents. They provide gels with enhanced resolution and clarity, low EEO and low background. Tested to be RNase, DNase, Protease and Endonuclease free. Supplied in 100g bottles.
Catalog No. 50-238-9573 Supplier Alkali Scientific Supplier No. A6601
ASI™ Agarose LM is a low melting point agarose ideal for preparative gels. The melting and gelling points are well below that of the DNA/RNA denaturation temperatures, allowing for recovery of nucleotides from the gel. Also useful for cloning tissue culture cells and viral plaque assays. All of our agarose is manufactured under a “green” process that eliminates the use of organic solvents. They provide gels with enhanced resolution and clarity, low EEO and low background. Tested to be RNase, DNase, Protease and Endonuclease free. Supplied in 100g bottles.