Learn More
Alkali Scientific Agarose, Low EEO, 500 G
SDP
Catalog No. 5000660817
Encompass_Preferred
Agarose, Low EEO, 500 G is a molecular biology grade agarose designed for gel electrophoresis. This agarose has low electroendosmosis (EEO), which minimizes background migration, making it perfect for high-resolution separation of biomolecules such as nucleic acids and proteins. The 500g pack size is ideal for large-scale applications, high-throughput laboratories, and facilities performing routine electrophoresis. The low EEO property ensures that bands remain sharp and distinct, providing precise separation for research in genomics, proteomics, and molecular biology. This agarose is widely used for applications requiring clarity and precision in electrophoresis experiments.
Catalog No. 50-006-60817 Supplier Alkali Scientific Supplier No. AGA010CP
Agarose, Low EEO, 500 G is a molecular biology grade agarose designed for gel electrophoresis. This agarose has low electroendosmosis (EEO), which minimizes background migration, making it perfect for high-resolution separation of biomolecules such as nucleic acids and proteins. The 500g pack size is ideal for large-scale applications, high-throughput laboratories, and facilities performing routine electrophoresis. The low EEO property ensures that bands remain sharp and distinct, providing precise separation for research in genomics, proteomics, and molecular biology. This agarose is widely used for applications requiring clarity and precision in electrophoresis experiments.