Learn More
Research Products International Corp Agarose, Low EEO, 50 Grams
SDP
Catalog No. 50213124
Encompass_Preferred
Molecular Biology Grade. High purity for preparative and analytical electrophoresis of nucleic acids >1000 bp.
Gelling Temperature: 34.5 -37.5°C
Sulfate: 0.15
EEO: 0.09- 0.13
DNA Binding: None Detected
DNA and RNase: None Detected
Catalog No. 50-213-124 Supplier Research Products International Corp Supplier No. A2008050.0
Molecular Biology Grade. High purity for preparative and analytical electrophoresis of nucleic acids >1000 bp.
Gelling Temperature: 34.5 -37.5°C
Sulfate: 0.15
EEO: 0.09- 0.13
DNA Binding: None Detected
DNA and RNase: None Detected