Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Agarose, Low Melt, 1 KG
SDP

Catalog No. 501976856
Encompass_Preferred

Ideal gelling properties for use in cloning assays or for recovery of heat labile samples following electrophoresis.

Gel Strength, 1%: >200 g/cm2
Gelling Temperature: 26-30°C
Melting Temperature: <65°C
Sulfate: <0.10%
EEO: <0.10
DNase and RNase: None Detected

Catalog No. 50-197-6856 Supplier Research Products International Corp Supplier No. A200701000.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ideal gelling properties for use in cloning assays or for recovery of heat labile samples following electrophoresis.

Gel Strength, 1%: >200 g/cm2
Gelling Temperature: 26-30°C
Melting Temperature: <65°C
Sulfate: <0.10%
EEO: <0.10
DNase and RNase: None Detected

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.