Learn More
Biotang Inc AGAROSE LOW MELT, 100 GM, P-730,CAS # 9012-36-6. Molecular biology grade, granulated powder, gel Strength:~250 g/cm2 gelling temperature: < 40 C(approximately 25-30 C) and melting temperature:70-100C
Catalog No. 50152329 Shop All BIOTANG Inc Products
Encompass_Preferred
AGAROSE LOW MELT, 100 GM, P-730,CAS # 9012-36-6. Molecular biology grade, granulated powder, gel Strength:~250 g/cm2 gelling temperature: < 40 C(approximately 25-30 C) and melting temperature:70-100C.
Catalog No. 50-152-329 Supplier Biotang Inc Supplier No. BTBB875
AGAROSE LOW MELT, 100 GM, P-730,CAS # 9012-36-6. Molecular biology grade, granulated powder, gel Strength:~250 g/cm2 gelling temperature: < 40 C(approximately 25-30 C) and melting temperature:70-100C.