Learn More
IBI SCIENTIFIC 3P Low Melt Agarose | 9012-36-6 | MFCD00081294 | 100 g
Catalog No. 50550460
Encompass_Preferred
Agarose, Low Melting Point - 100gm IBI Molecular Biology grade Low Melt Agarose is commonly used as a gel matrix when fragment recovery after electrophoresis is desired. It can also be used for preparing large DNA samples for plugs.
Catalog No. 50-550-460 Supplier IBI SCIENTIFIC 3P Supplier No. IB70056
Agarose, Low Melting Point - 100gm IBI Molecular Biology grade Low Melt Agarose is commonly used as a gel matrix when fragment recovery after electrophoresis is desired. It can also be used for preparing large DNA samples for plugs.