Learn More
IBI SCIENTIFIC 3P Low Melt Agarose | 9012-36-6 | MFCD00081294 | 25 g
Catalog No. 50550455
Encompass_Preferred
IBI Molecular Biology grade Low Melt Agarose is commonly used as a gel matrix when fragment recovery after electrophoresis is desired. It can also be used for preparing large DNA samples for plugs. Melting Tempurature at 1.5% gel is 65.5C.
Catalog No. 50-550-455 Supplier IBI SCIENTIFIC 3P Supplier No. IB70051
IBI Molecular Biology grade Low Melt Agarose is commonly used as a gel matrix when fragment recovery after electrophoresis is desired. It can also be used for preparing large DNA samples for plugs. Melting Tempurature at 1.5% gel is 65.5C.