Learn More
IBI SCIENTIFIC 3P AGAROSE LOW MELT POINT-50GM
Catalog No. 50550454
Encompass_Preferred
Agarose, Low Melting Point - 50gm IBI Molecular Biology grade Low Melt Agarose is commonly used as a gel matrix when fragment recovery after electrophoresis is desired. It can also be used for preparing large DNA samples for plugs
Catalog No. 50-550-454 Supplier IBI SCIENTIFIC 3P Supplier No. IB70050
Agarose, Low Melting Point - 50gm IBI Molecular Biology grade Low Melt Agarose is commonly used as a gel matrix when fragment recovery after electrophoresis is desired. It can also be used for preparing large DNA samples for plugs