Learn More
Alkali Scientific Agarose, Low Melt Temperature, 1 Kg
SDP
Supplier:  Alkali Scientific AGA002CP
Encompass_Preferred
Agarose, Low Melt Temperature, 1 Kg is a large-volume reagent used in molecular biology and biochemistry for gel electrophoresis, particularly in DNA and RNA analysis. This low-melt agarose provides superior gel formation and is ideal for procedures that require melting and recasting of gels, such as in pulsed-field gel electrophoresis. The 1 kg package is perfect for high-throughput laboratories that need large quantities of agarose for routine gel preparation. The low-melt feature allows for efficient handling and processing of gels without compromising resolution, ensuring that large DNA fragments are separated accurately during electrophoresis.
Catalog No. 50-006-60664