Learn More
Alkali Scientific Agarose, Low Melt Temperature, 100 G
SDP
Catalog No. 5000660154
Encompass_Preferred
Agarose, Low Melt Temperature, 100 G is a molecular biology-grade reagent used in gel electrophoresis applications. This low-melt agarose is designed to provide a flexible gel matrix that melts at a lower temperature, which is ideal for applications where the gel needs to be reheated or manipulated. It is widely used in DNA and RNA electrophoresis, as well as in other techniques like blotting, where the ability to easily dissolve and re-prepare the gel is necessary. The 100g package is suitable for laboratories with moderate throughput, offering high-quality gels that provide excellent resolution and reproducibility in molecular biology experiments.
Catalog No. 50-006-60154 Supplier Alkali Scientific Supplier No. AGA001CP
Agarose, Low Melt Temperature, 100 G is a molecular biology-grade reagent used in gel electrophoresis applications. This low-melt agarose is designed to provide a flexible gel matrix that melts at a lower temperature, which is ideal for applications where the gel needs to be reheated or manipulated. It is widely used in DNA and RNA electrophoresis, as well as in other techniques like blotting, where the ability to easily dissolve and re-prepare the gel is necessary. The 100g package is suitable for laboratories with moderate throughput, offering high-quality gels that provide excellent resolution and reproducibility in molecular biology experiments.