Learn More
LabScientific Agarose MB Grade-LE | 500g, general purpose, low EEO agarose for nucleic acids fragments with sizes ≥1,000bp. Agarose channels - drosophila larva.
Supplier:  LabScientific AGLE
Encompass
Our best selling, general purpose, low EEO agarose for nucleic acids fragments with sizes ≥1,000bp. One of our most versatile agaroses and can be used for the rapid separation of DNA and RNA fragments as well as PCR products, the preparation of plasmids, and for screening, cloning and blotting techniques. This product and many others are available with an optional Genetic Quality Tested (GQT) certificate recommended for DNA preparative gels.
Catalog No. NC1059072