Learn More
Research Products International Corp Agarose ME Medium EEO, Molecular Biology Grade, 100 Grams
SDP
Catalog No. 50488464
Encompass_Preferred
For DNA/RNA fragments >1000bp. Intermediate EEO.
Catalog No. 50-488-464 Supplier Research Products International Corp Supplier No. A20085100.0
For DNA/RNA fragments >1000bp. Intermediate EEO.