Learn More
Research Products International Corp Agarose ME Medium EEO, Molecular Biology Grade, 500 Grams
SDP
Catalog No. 50488465
Encompass_Preferred
For DNA/RNA fragments >1000bp. Intermediate EEO.
Catalog No. 50-488-465 Supplier Research Products International Corp Supplier No. A20085500.0
For DNA/RNA fragments >1000bp. Intermediate EEO.