Learn More
Sigma Aldrich Fine Chemicals Biosciences Agarose Medium EEO for mol100G
Catalog No. 502903525
Encompass
Agarose is a polymer extracted from agar or agar-bearing marine algae. This purified linear galactan hydrocolloid comprises alternating co-polymers D-galactose and 3 6-anhydro-L-galactose units connected by (1A†3) and -(1A†4) glycosidic bonds. Agarose is highly biocompatible possesses variable mechanical and diffusion properties.
Catalog No. 50-290-3525 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A6877100G
Agarose is a polymer extracted from agar or agar-bearing marine algae. This purified linear galactan hydrocolloid comprises alternating co-polymers D-galactose and 3 6-anhydro-L-galactose units connected by (1A†3) and -(1A†4) glycosidic bonds. Agarose is highly biocompatible possesses variable mechanical and diffusion properties.