Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals Agarose, Molecular Biology Grade

Catalog No. 11AGAH0100 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 x 100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11AGAH0100 1 x 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11AGAH0100 Supplier MP Biomedicals Supplier No. 11AGAH0100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Essential component in chemical separation techniques.

  • Standard agarose is a purified linear galactan hydrocolloid isolated from agar or agar-bearing marine algae with an Electroendoosmosis (EEO) value of 0.12 making it an essential component in chemical separation techniques.
  • Key applications: Electrophoresis, separation of DNA/RNA fragments from 400 to 8000 bp.
  • Research Use Only (RUO). Ready for CE IVD labeling of clinical applications.
TRUSTED_SUSTAINABILITY

Specifications

Description Typical working solution: 1.5% gel
Color White
For Use With (Application) Molecular biology, electrophoresis, separation of DNA/RNA fragments from 400 to 8000 bp
Melting Temperature 88°C ±1.5°C (1.5% gel)
Quantity 1 x 100 g
Storage Requirements Room temperature
Product Type Agarose gel powder
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.