Learn More
MP Biomedicals™ Agarose, MP Biomedicals™
Catalog No. MP210026780 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
Catalog No. Quantity
MP210026780 100 g
1 options
Catalog No. MP210026780 Supplier MP Biomedicals™ Supplier No. 0210026780
Add to Cart
Add to Cart
Suitable for both analytical and preparative electrophoresis procedures. Agarose, MP Biomedicals is a purified linear galactan hydrocolloid isolated from agar or agar-bearing algae. It is specially prepared to recover active DNA after separation, with min
This is a standard agarose with good gel strength and clarity, suitable for all general applications.
  • Useful in separation of nucleic acids electrophoretically, because agarose gels have larger pore sizes than cross linked acrylamide gels at low concentrations
  • Useful in chromatographic separations by either beading the agarose or cross-linking it
  • Agarose gels are used in immunoelectrophoresis (IEP) and double-diffusion Ouchterlony plates to demonstrate antibody-antigen reactions
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 g
Product Type Agarose Gel
WARNING: Cancer - www.P65Warnings.ca.gov