Learn More
Sigma Aldrich Fine Chemicals Biosciences Agarose MP multipurpose ag
Catalog No. 502902884
Encompass
Agarose is obtained from seaweed and contains repeated agarobiose units. During agarose gel formation the polymers aggregate to form a network with varying pore sizes. This property is used in agarose gel electrophoresis to separate DNA.
Catalog No. 50-290-2884 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11388991001
Agarose is obtained from seaweed and contains repeated agarobiose units. During agarose gel formation the polymers aggregate to form a network with varying pore sizes. This property is used in agarose gel electrophoresis to separate DNA.