Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agarose, PFGE, Ultrapure Bioreagent, J.T. Baker™

Catalog No. 02002206 Shop All Avantor Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-002-206 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 02-002-206 Supplier Avantor Supplier No. 406305
Add to Cart
Add to Cart
This item is not returnable. View return policy

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Rapid Resolution of DNA Greater than 1,000 bp

Specifications

CAS 9012-36-6
Quantity 100 g
Packaging Poly Bottle
Grade Ultrapure Bioreagent
Chemical Name or Material Agarose, PFGE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.