Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Agarose SFR™ Agarose S FR White, Fine Powder 100 GR
SDP

Catalog No. 50980168
Encompass_Preferred

CAS #9012-36-6 (C₆H₁₀O₅·C₆H₈O₄)n
Melting point: ≤ 70°C

Agarose SFR is a high resolution sieving agarose with unsurpassed clarity. DNA bands differing in size by 2% can be resolved in the range of 200 – 1,000 bp. This agarose is suitable for the analysis of AFLP's (Amplified Fragment Length Polymorphisms), STR's (Short Tandem Repeats) and tetranucleotide repeats. The low melting point of agarose SFR makes it an excellent medium for analytical and preparative electrophoresis.

Catalog No. 50-980-168 Supplier Electron Microscopy Sciences Supplier No. 10207
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS #9012-36-6 (C₆H₁₀O₅·C₆H₈O₄)n
Melting point: ≤ 70°C

Agarose SFR is a high resolution sieving agarose with unsurpassed clarity. DNA bands differing in size by 2% can be resolved in the range of 200 – 1,000 bp. This agarose is suitable for the analysis of AFLP's (Amplified Fragment Length Polymorphisms), STR's (Short Tandem Repeats) and tetranucleotide repeats. The low melting point of agarose SFR makes it an excellent medium for analytical and preparative electrophoresis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.