Learn More
Crescent Chemical Co Inc AGAROSE SERVA DNA ELECTRO.
SDP
Supplier:  Crescent Chemical Co Inc SE11404.07
Encompass_Preferred
Agarose SERVA for DNA electrophoresis research grade For analytical and preparative electrophoresis of DNA fragments between 1000 and 20,000 bp. Each lot is tested for the absence of EcoRI inhibition. Size - 500G Storage Conditions - +15 °C TO +30 °C Catalog Number - 11
Catalog No. 50-247-220