Learn More
Crescent Chemical Co Inc AGAROSE SERVA DNA ELECTRO.
SDP
Catalog No. 50247219
Encompass_Preferred
Agarose SERVA for DNA electrophoresis research grade For analytical and preparative electrophoresis of DNA fragments between 1000 and 20,000 bp. Each lot is tested for the absence of EcoRI inhibition. Size - 1KG Storage Conditions - +15 °C TO +30 °C Catalog Number - 114
Catalog No. 50-247-219 Supplier Crescent Chemical Co Inc Supplier No. SE11404.05
Agarose SERVA for DNA electrophoresis research grade For analytical and preparative electrophoresis of DNA fragments between 1000 and 20,000 bp. Each lot is tested for the absence of EcoRI inhibition. Size - 1KG Storage Conditions - +15 °C TO +30 °C Catalog Number - 114