Learn More
Crescent Chemical Co Inc AGAROSE SERVA LOW MELTING
SDP
Supplier:  Crescent Chemical Co Inc SE11408.01
Encompass_Preferred
Agarose SERVA low melting research grade For analytical and preparative nucleic acid electrophoresis of DNA/RNA fragments \> 500 bp. Highly efficient recovery of DNA fragments at low temperature for subsequent in-gel manipulations like restriction analysis or ligation reac
Catalog No. 50-247-226