Learn More
Crescent Chemical Co Inc AGAROSE SERVA LOW MELTING
SDP
Catalog No. 50247227
Encompass_Preferred
Agarose SERVA low melting research grade For analytical and preparative nucleic acid electrophoresis of DNA/RNA fragments \> 500 bp. Highly efficient recovery of DNA fragments at low temperature for subsequent in-gel manipulations like restriction analysis or ligation reac
Catalog No. 50-247-227 Supplier Crescent Chemical Co Inc Supplier No. SE11408.02
Agarose SERVA low melting research grade For analytical and preparative nucleic acid electrophoresis of DNA/RNA fragments \> 500 bp. Highly efficient recovery of DNA fragments at low temperature for subsequent in-gel manipulations like restriction analysis or ligation reac