Learn More
Crescent Chemical Co Inc AGAROSE SERVA NEUTRAL IEF
SDP
Catalog No. 50247214
Encompass_Preferred
Agarose SERVA neutral for IEF Premium grade for isoelectric focusing. Chemically treated agarose to neutralize residual negative charge sites, virtually eliminating electroendosmosis. Size - 5G Storage Conditions - +15 °C TO +30 °C Catalog Number - 11402.02
Catalog No. 50-247-214 Supplier Crescent Chemical Co Inc Supplier No. SE11402.02
Agarose SERVA neutral for IEF Premium grade for isoelectric focusing. Chemically treated agarose to neutralize residual negative charge sites, virtually eliminating electroendosmosis. Size - 5G Storage Conditions - +15 °C TO +30 °C Catalog Number - 11402.02