Learn More
Crescent Chemical Co Inc AGAROSE SERVA PCR GRADE
SDP
Catalog No. 50247192
Encompass_Preferred
Agarose SERVA for PCR molecular biology grade For analytical and preparative electrophoresis of PCR and DNA fragments \> 40 bp and \ Size - 100G Storage Conditions - +15 °C TO +30 °C Catalog Number - 11383.02 CAS 9012-36-6
Catalog No. 50-247-192 Supplier Crescent Chemical Co Inc Supplier No. SE11383.02
Agarose SERVA for PCR molecular biology grade For analytical and preparative electrophoresis of PCR and DNA fragments \> 40 bp and \ Size - 100G Storage Conditions - +15 °C TO +30 °C Catalog Number - 11383.02 CAS 9012-36-6