Learn More
Crescent Chemical Co Inc AGAROSE SERVA PCR LOW MELT
SDP
Catalog No. 50247194
Encompass_Preferred
Agarose SERVA for PCR low melting molecular biology grade For analytical and preparative electrophoresis of PCR and DNA fragments \> 40 bp and \ Size - 25G Storage Conditions - +15 °C TO +30 °C Catalog Number - 11384.01 CAS 9012-36-6
Catalog No. 50-247-194 Supplier Crescent Chemical Co Inc Supplier No. SE11384.01
Agarose SERVA for PCR low melting molecular biology grade For analytical and preparative electrophoresis of PCR and DNA fragments \> 40 bp and \ Size - 25G Storage Conditions - +15 °C TO +30 °C Catalog Number - 11384.01 CAS 9012-36-6