Learn More
Crescent Chemical Co Inc AGAROSE SERVA PREM MOL. BIO
SDP
Catalog No. 50247187
Encompass_Preferred
Agarose SERVA Premium molecular biology grade For analytical and preparative gel electrophoresis of DNA/RNA fragments \> 500 bp, recovery of DNA fragments for further modifications (restriction analysis, ligation reactions), blotting of nucleic acids. Special quality te
Catalog No. 50-247-187 Supplier Crescent Chemical Co Inc Supplier No. SE11381.03
Agarose SERVA Premium molecular biology grade For analytical and preparative gel electrophoresis of DNA/RNA fragments \> 500 bp, recovery of DNA fragments for further modifications (restriction analysis, ligation reactions), blotting of nucleic acids. Special quality te