Learn More
Crescent Chemical Co Inc AGAROSE SERVA RES. GRADE
SDP
Supplier:  Crescent Chemical Co Inc SE11380.02
Encompass_Preferred
Agarose SERVA research grade Agarose with low EEO for analytical and preparative gel electrophoresis and blotting of DNA/RNA fragments \> 500 bp. Size - 100G Storage Conditions - +15 °C TO +30 °C Catalog Number - 11380.02 CAS 9012-36-6
Catalog No. 50-247-182