Learn More
Crescent Chemical Co Inc AGAROSE SERVA RES. GRADE
SDP
Catalog No. 50247183
Encompass_Preferred
Agarose SERVA research grade Agarose with low EEO for analytical and preparative gel electrophoresis and blotting of DNA/RNA fragments \> 500 bp. Size - 250G Storage Conditions - +15 °C TO +30 °C Catalog Number - 11380.03 CAS 9012-36-6
Catalog No. 50-247-183 Supplier Crescent Chemical Co Inc Supplier No. SE11380.03
Agarose SERVA research grade Agarose with low EEO for analytical and preparative gel electrophoresis and blotting of DNA/RNA fragments \> 500 bp. Size - 250G Storage Conditions - +15 °C TO +30 °C Catalog Number - 11380.03 CAS 9012-36-6