Learn More
Crescent Chemical Co Inc AGAROSE SERVA WIDE RANGE
SDP
Supplier:  Crescent Chemical Co Inc SE11406.02
Encompass_Preferred
Agarose SERVA Wide Range molecular biology grade For analytical and preparative electrophoresis and blotting of DNA/RNA fragments between 250 and 23,000 bp, PCR products, preparation of plasmids, screening and cleaning. Tested for applications in molecular biology. Si
Catalog No. 50-247-224