Learn More
Crescent Chemical Co Inc AGAROSE SERVA WIDE RANGE
SDP
Catalog No. 50247225
Encompass_Preferred
Agarose SERVA Wide Range molecular biology grade For analytical and preparative electrophoresis and blotting of DNA/RNA fragments between 250 and 23,000 bp, PCR products, preparation of plasmids, screening and cleaning. Tested for applications in molecular biology. Si
Catalog No. 50-247-225 Supplier Crescent Chemical Co Inc Supplier No. SE11406.03
Agarose SERVA Wide Range molecular biology grade For analytical and preparative electrophoresis and blotting of DNA/RNA fragments between 250 and 23,000 bp, PCR products, preparation of plasmids, screening and cleaning. Tested for applications in molecular biology. Si