Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Agarose Standard

Catalog No. MP1AGAH0250 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP1AGAH0250 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP1AGAH0250 Supplier MP Biomedicals™ Supplier No. 11AGAH0250
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Agarose Standard, Molecular Biology Grade, White Powder

  • Standard agarose for analytical electrophoresis
  • Low EEO for complete resolution of bands >500 bp
  • Highly purifi ed, strict quality test procedures
  • Tested for transfer to membrane
  • QA-AgaroseTM Standard is a standard gelling temperature agarose with very low EEO (electroendosmosis) values giving complete resolution
  • of bands greater than 500 bp.
  • This agarose is recommended for analysis when sharp resolution of bands >500 bp is required.
  • The product is quality tested on a lot basis to ensure the absence of DNases and RNases and for the absence of inhibitors to restriction enzymes.
  • Each batch of agarose must pass a DNA transfer test whereby full DNA transfer must occur from gel to membrane in a 1 hour vacuum transfer procedure.
TRUSTED_SUSTAINABILITY

Specifications

CAS 9012-36-6
Description Standard agarose is a purified lineargalactan hydrocolloid isolated from agar or agar-bearing marine algae with an Electroendoosmosis (EEO) value of 0.12 making it an essential component in chemical separation techniques.
Color White
Form Powder
For Use With (Application) Electrophoresis; Separation of DNA/RNA fragments from 400 to 8000 bp
Melting Temperature 89°C (1.5%gel)
Quantity 250 g
Storage Requirements 15 - 30°C
Product Type Agarose Gel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.