Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Agarose 500mg Tablets, 200 Tablets (100 Grams)
SDP

Catalog No. 50488469
Encompass_Preferred

Save time and labor preparing agarose gels for electrophoresis of nucleic acids. Just place the required number of tablets into a standard buffer, then mix and heat until completely dissolved.

Tablet Weight Range: 485-515 mg
Gel Strength, 1.5%: >1200 g/cm2
Gelling Range: 36-39°C
Melting Temp: 87-89°C
EEO: <0.10
Sulfate: <0.15
DNase, RNase: None Detected

Catalog No. 50-488-469 Supplier Research Products International Corp Supplier No. A20096100.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Save time and labor preparing agarose gels for electrophoresis of nucleic acids. Just place the required number of tablets into a standard buffer, then mix and heat until completely dissolved.

Tablet Weight Range: 485-515 mg
Gel Strength, 1.5%: >1200 g/cm2
Gelling Range: 36-39°C
Melting Temp: 87-89°C
EEO: <0.10
Sulfate: <0.15
DNase, RNase: None Detected

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.