Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biotang Inc Agarose Blenders - TAE Blend 0.8%, 500 ml

Catalog No. 50152321 Shop All BIOTANG Inc Products
Encompass_Preferred

Agarose Blenders - TAE Blend 0.8%, 500 ml.

Catalog No. 50-152-321 Supplier Biotang Inc Supplier No. BTBB868
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Agarose Blenders - TAE Blend 0.8%, 500 ml.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.