Learn More
Embi Tec AGAROSE TBE W/ETBR 12+1 WELL
SDP
Catalog No. NC9225868
Encompass
RunOne Pre-Castgel, 1% agarose TBE w/ETBR landscapegel 12+1 well, load volume 30uL, optimal separation 600 to 10000 bp 10/PK
Catalog No. NC9225868 Supplier Embi Tec Supplier No. GE4600
RunOne Pre-Castgel, 1% agarose TBE w/ETBR landscapegel 12+1 well, load volume 30uL, optimal separation 600 to 10000 bp 10/PK