Learn More
LifeSensors Tandem Ubiquitin Binding Entities (TUBE1) Agarose 1mL
SDP
Supplier:  LifeSensors UM04011000
Encompass_Preferred
Based on protein domains known to possess an affinity for ubiquitin, Tandem Ubiquitin Binding Entities (TUBEs) have been developed for the isolation and identification of ubiquitinated proteins. TUBEs display up to a 1000-fold increase in affinity for polyubiquitin moieties over the single ubiquitin binding associated domain (UBA). In addition, TUBEs protect polyubiquitinated proteins from deubiquitination and proteasomal degradation, allowing for detection at relatively low abundance. These properties effectively allow TUBEs to “capture” protein in its polyubiqutinated state. TUBEs can be used to isolate, enrich, and identify ubiquitinated proteins from cell and tissue extracts. Agarose coupled TUBEs allow for efficient recovery of polyubiquitinated proteins in a single step. Polyubiquitinated protein can easily be separated from TUBEs for proteomic studies or identification by Western blotting. This product is protected by one or more US or Foreign patents.
Catalog No. 50-103-7771