Learn More
Alkali Scientific Agarose UHR, Ultra High Resolution, 100 G
SDP
Supplier:  Alkali Scientific SEU001CP
Encompass_Preferred
Ultra High Resolution Agarose is a premium agarose used in DNA gel electrophoresis to achieve the highest resolution in DNA fragment separation. This 100g package is perfect for applications requiring the most precise separation, such as separating very small or closely related DNA fragments. Ultra High Resolution Agarose is ideal for use in high-performance applications, where precise band separation and clear visualization are essential. It is commonly used in laboratories for the analysis of PCR products, restriction enzyme digests, and other genomic studies where resolving small differences in DNA sizes is critical.
Catalog No. 50-006-61617