Learn More
Alkali Scientific Agarose UHR, Ultra High Resolution, 500 G
SDP
Catalog No. 5000661081
Encompass_Preferred
Ultra High Resolution Agarose in a 500g package is designed for high-performance DNA gel electrophoresis. This large quantity is ideal for labs that require frequent, high-quality gel electrophoresis, particularly for separating very small or complex DNA fragments. The agarose's high resolution provides sharp band separation and high clarity, making it perfect for research involving small PCR products, DNA sequencing, or genetic fingerprinting. This agarose ensures reproducible and accurate results, and is an essential reagent for labs performing advanced genetic analysis, molecular biology, and other high-resolution DNA separation applications.
Catalog No. 50-006-61081 Supplier Alkali Scientific Supplier No. SEU003CP
Ultra High Resolution Agarose in a 500g package is designed for high-performance DNA gel electrophoresis. This large quantity is ideal for labs that require frequent, high-quality gel electrophoresis, particularly for separating very small or complex DNA fragments. The agarose's high resolution provides sharp band separation and high clarity, making it perfect for research involving small PCR products, DNA sequencing, or genetic fingerprinting. This agarose ensures reproducible and accurate results, and is an essential reagent for labs performing advanced genetic analysis, molecular biology, and other high-resolution DNA separation applications.