Learn More
Sigma Aldrich Fine Chemicals Biosciences Agate mortar and pestle St
Catalog No. 502905311
Encompass
Made of natural agate with highly polished grinding surfaces. Excellent for preparing laboratory samples and high purity powders. The agate material is a special silicate with a main composition of SiO2. It is resistant to detergent. The thermal endurance is limited up to 350C.
Catalog No. 50-290-5311 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z1125181SET
Made of natural agate with highly polished grinding surfaces. Excellent for preparing laboratory samples and high purity powders. The agate material is a special silicate with a main composition of SiO2. It is resistant to detergent. The thermal endurance is limited up to 350C.