Learn More
Chemplex Industries AGATE MORTAR & PESTLE 1.97IN
Catalog No. 50249473
Encompass_Preferred
Agate Mortar and Pestle Sets, 1.97 (50 mm). Natural quartz; Highly resistant to wear and abrasion; Extremely hard - 6 on the Mohs scale.
Catalog No. 50-249-473 Supplier Chemplex Industries Supplier No. 8050
Agate Mortar and Pestle Sets, 1.97 (50 mm). Natural quartz; Highly resistant to wear and abrasion; Extremely hard - 6 on the Mohs scale.