Learn More
Medchemexpress LLC Agatolimod sodium | 541547-35-7 | DNA, d(P-thio)(T-C-G-T-C-G-T-T-T-T-G-T-C-G-T-T-T-T-G-T-C-G-T-T), tricosasodium salt | 1 MG

Supplier: Medchemexpress LLC HY1502181MG
Agatolimod sodium (ODN 2006) is a class B CpG ODN and acts as a TLR9 agonist. It can be utilized as a vaccine adjuvant and is applicable for cancer research.
- Class B CpG ODN
- TLR9 agonist
- Vaccine adjuvant
- Applicable for cancer research
- Sequence: 5'-TCGTCGTTTTGTCGTTTTGTCGTT-3'
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.