Learn More
Medchemexpress LLC ω-Agatoxin IVA TFA | 99.1% | 5316.27 | 1 MG
SDP
Catalog No. 5000423028
Encompass_Preferred
ω-Agatoxin IVA TFA is a potent and selective blocker of P/Q-type Ca2+ (Cav2.1) channels. It inhibits glutamate exocytosis and calcium influx induced by high potassium, and blocks the high potassium-induced release of serotonin and norepinephrine. This compound does not affect L-type or N-type calcium channels.
  • Potent and selective blocker of P/Q-type Ca2+ (Cav2.1) channels
  • Inhibits glutamate exocytosis and calcium influx induced by high potassium
  • Blocks high potassium-induced release of serotonin and norepinephrine
  • Does not affect L-type or N-type calcium channels
Catalog No. 50-004-23028 Supplier Medchemexpress LLC Supplier No. HYP1080A1MG
ω-Agatoxin IVA TFA is a potent and selective blocker of P/Q-type Ca2+ (Cav2.1) channels. It inhibits glutamate exocytosis and calcium influx induced by high potassium, and blocks the high potassium-induced release of serotonin and norepinephrine. This compound does not affect L-type or N-type calcium channels.
  • Potent and selective blocker of P/Q-type Ca2+ (Cav2.1) channels
  • Inhibits glutamate exocytosis and calcium influx induced by high potassium
  • Blocks high potassium-induced release of serotonin and norepinephrine
  • Does not affect L-type or N-type calcium channels