Learn More
Medchemexpress LLC ω-Agatoxin IVA TFA | 99.1% | 5316.27 | 100 UG
SDP
Catalog No. 5000423027
Encompass_Preferred
ω-Agatoxin IVA TFA is a potent, selective P/Q type Ca2+ (Cav2.1) channel blocker. It has IC50s of 2 nM for P-type and 90 nM for Q-type Ca2+ channels. It inhibits glutamate exocytosis and calcium influx elicited by high potassium (IC50, 30-225 nM), and blocks the high potassium-induced release of serotonin and norepinephrine. It has no effect on L-type or N-type calcium channels.
  • Potent and selective P/Q type Ca2+ (Cav2.1) channel blocker
  • Inhibits glutamate exocytosis and calcium influx
  • Blocks high potassium-induced release of serotonin and norepinephrine
  • Does not affect L-type or N-type calcium channels
Catalog No. 50-004-23027 Supplier Medchemexpress LLC Supplier No. HYP1080A100UG
ω-Agatoxin IVA TFA is a potent, selective P/Q type Ca2+ (Cav2.1) channel blocker. It has IC50s of 2 nM for P-type and 90 nM for Q-type Ca2+ channels. It inhibits glutamate exocytosis and calcium influx elicited by high potassium (IC50, 30-225 nM), and blocks the high potassium-induced release of serotonin and norepinephrine. It has no effect on L-type or N-type calcium channels.
  • Potent and selective P/Q type Ca2+ (Cav2.1) channel blocker
  • Inhibits glutamate exocytosis and calcium influx
  • Blocks high potassium-induced release of serotonin and norepinephrine
  • Does not affect L-type or N-type calcium channels