Learn More
Medchemexpress LLC AgAuSe-PEG-NH2 (1000 nm) | 250 UL
SDP
Catalog No. 5000429322
Encompass_Preferred
AgAuSe-PEG-NH2 (1000 nm) is a fluorescent quantum dot that emits fluorescence in the near-infrared II region, with an emission peak reaching 1000 nm that can be used in bioimaging.
  • Emits fluorescence in the near-infrared II region
  • Emission peak reaching 1000 nm
  • Useful for bioimaging
Catalog No. 50-004-29322 Supplier Medchemexpress LLC Supplier No. 250ULHYD2195
AgAuSe-PEG-NH2 (1000 nm) is a fluorescent quantum dot that emits fluorescence in the near-infrared II region, with an emission peak reaching 1000 nm that can be used in bioimaging.
  • Emits fluorescence in the near-infrared II region
  • Emission peak reaching 1000 nm
  • Useful for bioimaging