Learn More
Adam Equipment AGB 260a - AGB and AGF Bench and Floor Scales - Capacity: 260lb / 120kg, Readability: 0.01lb / 5g
Catalog No. NC3964928
Encompass_Preferred
AGB and AGF bench and floor scales from Adam Equipment deliver precision and durability for demanding industrial environments. Built with rugged construction and an IP68-rated stainless steel indicator, they withstand washdowns and harsh conditions. Available in multiple platform sizes and capacities, they support weighing, parts counting, percentage weighing, and checkweighing, with digital filtering for accurate results on moving items.A large backlit LCD with 40mm digits ensures clear visibility, while a color-changing display and color-coded keys improve efficiency. Two hold functions—display and peak—allow easy weight capture. A rechargeable battery provides up to 120 hours of operation, ideal for portable use.With RS-232 connectivity, programmable auto power-off, external calibration, and preset tare, AGB and AGF scales offer reliable, efficient performance for industrial applications.
Catalog No. NC3964928 Supplier Adam Equipment Supplier No. AGB 260A
AGB and AGF bench and floor scales from Adam Equipment deliver precision and durability for demanding industrial environments. Built with rugged construction and an IP68-rated stainless steel indicator, they withstand washdowns and harsh conditions. Available in multiple platform sizes and capacities, they support weighing, parts counting, percentage weighing, and checkweighing, with digital filtering for accurate results on moving items.A large backlit LCD with 40mm digits ensures clear visibility, while a color-changing display and color-coded keys improve efficiency. Two hold functions—display and peak—allow easy weight capture. A rechargeable battery provides up to 120 hours of operation, ideal for portable use.With RS-232 connectivity, programmable auto power-off, external calibration, and preset tare, AGB and AGF scales offer reliable, efficient performance for industrial applications.