Learn More
Aviva Systems Biology Corp AGBL5 Antibody - C-terminal region (ARP53752_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53752_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AGBL5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human AGBL5. Alternate names: FLJ21839ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-61116