Learn More
Grainger Fuse, Glass Fuse, Fast Acting, 1A, AGC Series, 5/pk
Supplier:  Grainger 4XH40
Encompass_Preferred
These cartridge-style, fast-acting glass fuses open within seconds of being exposed to overloads and short circuits. They're commonly used in instrument panels, small appliance circuits, and automotive accessories.
Catalog No. NC1701393