Learn More
MSC NES Drop Indicator Contact Point Extension, 1-3/4 in, #4-48 Thread, AGD
Catalog No. 5000516507
Encompass
This drop indicator contact point extension is designed for precise measurements with AGD dial indicators. It features a 1-3/4 inch length and a #4-48 thread, making it suitable for a variety of testing applications.
  • Provides 1-3/4 inch extension for dial indicators.
  • Features a #4-48 thread size.
  • Compatible with AGD dial indicators.
  • Manufactured in the United States.
  • Constructed from steel for durability.
Catalog No. 50-005-16507 Supplier MSC Supplier No. 01747716
This drop indicator contact point extension is designed for precise measurements with AGD dial indicators. It features a 1-3/4 inch length and a #4-48 thread, making it suitable for a variety of testing applications.
  • Provides 1-3/4 inch extension for dial indicators.
  • Features a #4-48 thread size.
  • Compatible with AGD dial indicators.
  • Manufactured in the United States.
  • Constructed from steel for durability.