Learn More
Abcam AGE-BSA, 10MG
Supplier:  Abcam AB5199510MG
Encompass_Preferred
AGE-BSA is modified bovine serum albumin (BSA) to create Advanced glycation end products (AGE)-BSA with >98% purity and suitable for SDS-PAGE. The predicted molecular weight of ab51995 protein is 66.5 kD. - Save time and ensure accurate and specific results by using our AGE- BSA protein as a control - Fluorescence of AGE-BSA: excitation/emission = 370/440 nm - Cited in over 20 publications
Protein/ Peptide/ Enzyme Synonyms: AGEs, Advanced glycation end products
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2148713