Learn More
Cayman Chemical AGEGAPyrIdIn SpecIfIc Mo 100ug
SDP
Catalog No. 502938858
Encompass_Preferred
Immunogen: GA-pyridine-BSA • Host: Mouse • Cross Reactivity: (-) (Carboxyethyl)-lysine (CEL), Glyoxal (GO), Methyl-glyoxal (MG) • Species reactivity: (+) Human • Applications: WB, ELISA
Catalog No. 50-293-8858 Supplier Cayman Chemical Supplier No. 27271100 UG
Immunogen: GA-pyridine-BSA • Host: Mouse • Cross Reactivity: (-) (Carboxyethyl)-lysine (CEL), Glyoxal (GO), Methyl-glyoxal (MG) • Species reactivity: (+) Human • Applications: WB, ELISA